Assay Details
| Gene Symbol: Ppm1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026447.4 | NP_080723.3 | 6 - 7 | 961 | 99 | |
| NM_198931.3 | NP_945149.2 | 6 - 7 | 965 | 99 | ||
| GenBank mRNA | AB474373.1 | - | 6 - 7 | 1000 | 99 | |
| AK013149.1 | - | 6 - 7 | 894 | 99 | ||
| AK017245.1 | - | 3 - 4 | 298 | 99 | ||
| AK046387.1 | - | 860 | 99 | |||
| AK087999.1 | - | 6 - 7 | 999 | 99 | ||
| AK216174.1 | - | 1 - 2 | 63 | 99 | ||
| AY332616.1 | - | 6 - 7 | 968 | 99 | ||
| BC058248.1 | - | 6 - 7 | 887 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppm1m
Genomic Map