Assay Details
| Gene Symbol: Ppm1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026447.4 | NP_080723.3 | 7 - 8 | 1022 | 118 | |
| NM_198931.3 | NP_945149.2 | 7 - 8 | 1026 | 118 | ||
| GenBank mRNA | AB474373.1 | - | 7 - 8 | 1061 | 118 | |
| AK013149.1 | - | 7 - 8 | 955 | 118 | ||
| AK017245.1 | - | 4 - 5 | 359 | 118 | ||
| AK046387.1 | - | 921 | 118 | |||
| AK087999.1 | - | 7 - 8 | 1060 | 118 | ||
| AK216174.1 | - | 2 - 3 | 124 | 118 | ||
| AY332616.1 | - | 7 - 8 | 1029 | 118 | ||
| BC058248.1 | - | 7 - 8 | 948 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppm1m
Genomic Map