Assay Details
| Gene Symbol: Ppm1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB474373.1 | - | 8 - 9 | 1171 | 60 | |
| AK087999.1 | - | 8 - 9 | 1170 | 60 | ||
| AY332616.1 | - | 8 - 9 | 1139 | 60 | ||
More Information
Set Membership: |
|

Ppm1m
Genomic Map