Assay Details
| Gene Symbol: Col6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102607.1 | NP_001096077.1 | 10 - 11 | 4187 | 93 | |
| GenBank mRNA | AK087828.1 | - | 10 - 11 | 4263 | 93 | |
| AM748261.1 | - | 3 - 4 | 1003 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Col6a6
Genomic Map