Assay Details
| Gene Symbol: Col6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102607.1 | NP_001096077.1 | 3 - 4 | 877 | 66 | |
| NM_172927.3 | NP_766515.2 | 3 - 4 | 912 | 66 | ||
| GenBank mRNA | AK054356.1 | - | 3 - 4 | 913 | 66 | |
| AK087828.1 | - | 3 - 4 | 953 | 66 | ||
| AM748260.1 | - | 1 - 2 | 597 | 66 | ||
| BC150695.1 | - | 2 - 3 | 696 | 9087606 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Col6a6
Genomic Map