Assay Details
| Gene Symbol: Slc16a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009197.2 | NP_033223.2 | 3 - 4 | 1231 | 79 | |
| GenBank mRNA | AB265788.1 | - | 3 - 4 | 1057 | 79 | |
| AF045692.1 | - | 3 - 4 | 1279 | 79 | ||
| BC080678.1 | - | 3 - 4 | 1067 | 6313495 | 79 | |
| BC096587.1 | - | 3 - 4 | 1167 | 3992886 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc16a2
Genomic Map