Assay Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF376726.1 | - | 13 - 14 | 1357 | 52 | |
| AK019012.1 | - | 4 - 5 | 331 | 52 | ||
| AK047364.1 | - | 13 - 14 | 1333 | 52 | ||
| AK082797.1 | - | 13 - 14 | 1382 | 52 | ||
| AK159313.1 | - | 13 - 14 | 1402 | 52 | ||
| AK170948.1 | - | 12 - 13 | 1485 | 52 | ||
| AK171030.1 | - | 12 - 13 | 1485 | 52 | ||
| AX775895.1 | - | 13 - 14 | 1414 | 52 | ||
| AX775897.1 | - | 13 - 14 | 1357 | 52 | ||
| BC016104.1 | - | 13 - 14 | 1383 | 4920840 | 52 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019012.1 transcripts.
Set Membership: |
|

Gpr108
Genomic Map