Assay Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF376726.1 | - | 15 - 16 | 1486 | 69 | |
| AK019012.1 | - | 6 - 7 | 460 | 69 | ||
| AK047364.1 | - | 15 - 16 | 1462 | 69 | ||
| AK082797.1 | - | 15 - 16 | 1511 | 69 | ||
| AK159313.1 | - | 15 - 16 | 1531 | 69 | ||
| AK170948.1 | - | 14 - 15 | 1614 | 69 | ||
| AK171030.1 | - | 14 - 15 | 1614 | 69 | ||
| AX775895.1 | - | 15 - 16 | 1543 | 69 | ||
| AX775897.1 | - | 15 - 16 | 1486 | 69 | ||
| BC016104.1 | - | 15 - 16 | 1512 | 4920840 | 69 | |
| BE981210.1 | - | 2 - 3 | 69 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr108
Genomic Map