Assay Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF376726.1 | - | 5 - 6 | 501 | 97 | |
| AK047364.1 | - | 5 - 6 | 477 | 97 | ||
| AK170948.1 | - | 5 - 6 | 529 | 97 | ||
| AK171030.1 | - | 5 - 6 | 529 | 97 | ||
| AX775897.1 | - | 5 - 6 | 501 | 97 | ||
| BG079632.2 | - | 5 - 6 | 541 | 97 | ||
More Information
Set Membership: |
|

Gpr108
Genomic Map