Assay Details
| Gene Symbol: Ranbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252466.1 | NP_001239395.1 | 9 - 10 | 952 | 85 | |
| NM_001252467.1 | NP_001239396.1 | 6 - 7 | 592 | 85 | ||
| NM_027933.3 | NP_082209.1 | 8 - 9 | 751 | 85 | ||
| NR_045519.1 | 9 - 10 | 861 | 85 | |||
| GenBank mRNA | AK011541.1 | - | 1 - 2 | 112 | 85 | |
| AK040481.1 | - | 10 - 11 | 935 | 85 | ||
| AK133949.1 | - | 8 - 9 | 598 | 85 | ||
| AK154201.1 | - | 7 - 8 | 2155 | 85 | ||
| AK155301.1 | - | 8 - 9 | 620 | 85 | ||
| AK167731.1 | - | 8 - 9 | 601 | 85 | ||
| BC066005.1 | - | 8 - 9 | 592 | 85 | ||
| BC145892.1 | - | 8 - 9 | 615 | 40131195 | 85 | |
| BC145894.1 | - | 8 - 9 | 615 | 40131197 | 85 | |
| CN717165.1 | - | 6 - 7 | 523 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ranbp3
Genomic Map