Assay Details
| Gene Symbol: Ranbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252466.1 | NP_001239395.1 | 15 - 16 | 1613 | 70 | |
| NM_001252467.1 | NP_001239396.1 | 12 - 13 | 1253 | 70 | ||
| NM_027933.3 | NP_082209.1 | 14 - 15 | 1412 | 70 | ||
| NR_045519.1 | 15 - 16 | 1522 | 70 | |||
| GenBank mRNA | AK011541.1 | - | 7 - 8 | 773 | 70 | |
| AK133949.1 | - | 14 - 15 | 1259 | 70 | ||
| AK134696.1 | - | 1 - 2 | 133 | 70 | ||
| AK155301.1 | - | 14 - 15 | 1281 | 70 | ||
| AK167731.1 | - | 14 - 15 | 1262 | 70 | ||
| BC066005.1 | - | 14 - 15 | 1253 | 70 | ||
| BC145892.1 | - | 14 - 15 | 1276 | 40131195 | 70 | |
| BC145894.1 | - | 14 - 15 | 1276 | 40131197 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ranbp3
Genomic Map