Assay Details
| Gene Symbol: Ranbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252466.1 | NP_001239395.1 | 5 - 6 | 553 | 85 | |
| NM_027933.3 | NP_082209.1 | 4 - 5 | 352 | 85 | ||
| GenBank mRNA | AK040481.1 | - | 6 - 7 | 536 | 85 | |
| AK133949.1 | - | 4 - 5 | 199 | 85 | ||
| AK154201.1 | - | 4 - 5 | 211 | 85 | ||
| AK155301.1 | - | 4 - 5 | 221 | 85 | ||
| AK167731.1 | - | 4 - 5 | 202 | 85 | ||
| BC066005.1 | - | 4 - 5 | 193 | 85 | ||
| BC145892.1 | - | 4 - 5 | 216 | 40131195 | 85 | |
| BC145894.1 | - | 4 - 5 | 216 | 40131197 | 85 | |
| CA318306.1 | - | 5 - 6 | 388 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ranbp3
Genomic Map