Assay Details
| Gene Symbol: Snx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110311.1 | NP_001103781.1 | 5 - 6 | 687 | 91 | |
| GenBank mRNA | AK131838.1 | - | 5 - 6 | 677 | 91 | |
| AK169569.1 | - | 5 - 6 | 663 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snx12
Genomic Map