Assay Details
| Gene Symbol: Xist | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001463.3 | 5 - 6 | 10101 | 81 | ||
| NR_001570.2 | 5 - 6 | 10101 | 81 | |||
| GenBank mRNA | L04961.1 | - | 5 - 6 | 10065 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis specific X-linked gene | Tsx | AV207184 | -44.2kb | Anti-sense | Chr. X - 103409101 - 103424588 on NCBI Build GRCm38 |
| Downstream |
Jpx transcript, Xist activator (non-protein coding) |
Jpx |
2010000I03Rik, 2510040I06Rik, Enox |
24.1kb | Anti-sense | Chr. X - 103493576 - 103506425 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Xist
Genomic Map