Assay Details
| Gene Symbol: Tmem263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013028.2 | NP_001013046.1 | 1 - 2 | 372 | 158 | |
| GenBank mRNA | AK005698.1 | - | 257 | 158 | ||
| AK142035.1 | - | 1 - 2 | 225 | 158 | ||
| BC086492.1 | - | 1 - 2 | 237 | 30470653 | 158 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005698.1, AK142035.1, BC086492.1, NM_001013028.2 transcripts.
Set Membership: |
|

Tmem263
Genomic Map