Assay Details
| Gene Symbol: Ppp1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028658.4 | NP_082934.3 | 2 - 3 | 263 | 94 | |
| GenBank mRNA | AK028185.1 | - | 2 - 3 | 127 | 94 | |
| AK155094.1 | - | 2 - 3 | 95 | 94 | ||
| AK170254.1 | - | 2 - 3 | 151 | 94 | ||
| BC106118.1 | - | 2 - 3 | 129 | 94 | ||
| BC137673.1 | - | 2 - 3 | 203 | 8860692 | 94 | |
| BY205567.1 | - | 2 - 3 | 263 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r21
Genomic Map