Assay Details
| Gene Symbol: Six3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015386.2 | 1 - 2 | 342 | 68 | ||
| NR_038082.1 | 1 - 2 | 342 | 68 | |||
| NR_038083.1 | 1 - 2 | 342 | 68 | |||
| NR_038084.1 | 1 - 2 | 342 | 68 | |||
| NR_038085.1 | 1 - 2 | 342 | 68 | |||
| NR_038086.1 | 1 - 2 | 342 | 68 | |||
| GenBank mRNA | AK053722.1 | - | 1 - 2 | 344 | 68 | |
| AY589790.1 | - | 1 - 2 | 221 | 68 | ||
| AY589792.1 | - | 1357 | 68 | |||
| BC065087.1 | - | 1 - 2 | 331 | 68 | ||
| CA527570.1 | - | 1 - 2 | 286 | 68 | ||
| CF737515.1 | - | 1 - 2 | 302 | 68 | ||
| CF737925.1 | - | 1 - 2 | 318 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calmodulin-lysine N-methyltransferase | Camkmt |
1700106N22Rik, AI480743, AV099781, CLNMT, CaM KMT |
-157.0kb | Anti-sense | Chr. 17 - 85090592 - 85458589 on NCBI Build GRCm38 |
| Downstream | sine oculis-related homeobox 3 | Six3 |
E130112M24Rik, Six3a, Six3alpha, Six3b, Six3beta |
2.5kb | Anti-sense | Chr. 17 - 85620834 - 85626191 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Six3os1
Genomic Map