Assay Details
| Gene Symbol: Six3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015386.2 | 2 - 3 | 485 | 96 | ||
| GenBank mRNA | AY589790.1 | - | 2 - 3 | 364 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calmodulin-lysine N-methyltransferase | Camkmt |
1700106N22Rik, AI480743, AV099781, CLNMT, CaM KMT |
-152.7kb | Anti-sense | Chr. 17 - 85090592 - 85458589 on NCBI Build GRCm38 |
| Downstream | sine oculis-related homeobox 3 | Six3 |
E130112M24Rik, Six3a, Six3alpha, Six3b, Six3beta |
5.1kb | Anti-sense | Chr. 17 - 85620834 - 85626191 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Six3os1
Genomic Map