Assay Details
| Gene Symbol: Vps51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081041.1 | NP_001074510.1 | 2 - 3 | 366 | 98 | |
| GenBank mRNA | AK137155.1 | - | 2 - 3 | 376 | 98 | |
| AK146211.1 | - | 2 - 3 | 375 | 98 | ||
| BC034452.1 | - | 2 - 3 | 382 | 98 | ||
| BC118930.2 | - | 2 - 3 | 381 | 40090408 | 98 | |
| BC118931.1 | - | 2 - 3 | 381 | 40090409 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041634 | NM_001081041.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269049 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69682 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544820 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278873 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5941 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119409 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041634 | NM_001081041.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03544102 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03544110 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119409 | - | - |
| Whole Mouse Genome G4122A | A_51_P340604 | AK003717.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14185328 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465358 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465362 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465359 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465355 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465356 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465363 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465360 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465361 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429834_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454825_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438205_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465354 | - | - |
| Mouse Genome 430 2.0 Array | 1438205_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429834_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1429834_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1454825_at | - | - |
| Mouse Genome 430 2.0 Array | 1438205_at | - | - |
| Mouse Genome 430 2.0 Array | 1454825_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68505_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68505_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900009203.mm.1 | NM_001081041.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009203.mm.1 | NM_001081041.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009203.mm.1 | TC1900001765.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454825_PM_at | - | - |
| Mouse Expression Set 430 | 1438205_PM_at | - | - |
| Mouse Expression Set 430 | 1454825_at | - | - |
| Mouse Expression Set 430 | 1438205_at | - | - |
| Murine Genome U74v2 Set | 96839_at | - | - |
| Murine Genome U74v2 Set | 96839_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429834_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429834_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438205_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438205_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454825_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454825_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 374792 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2570204 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2737979 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2607450 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2430830 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2607450 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2737979 | - | - |

Vps51
Genomic Map