Assay Details
| Gene Symbol: Rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009045.4 | NP_033071.1 | 4 - 5 | 681 | 90 | |
| GenBank mRNA | AK089489.1 | - | 4 - 5 | 426 | 90 | |
| AK144565.1 | - | 4 - 5 | 426 | 90 | ||
| AK154630.1 | - | 4 - 5 | 426 | 90 | ||
| AK155321.1 | - | 4 - 5 | 692 | 90 | ||
| AK155889.1 | - | 4 - 5 | 423 | 90 | ||
| AK157076.1 | - | 4 - 5 | 413 | 90 | ||
| AK158072.1 | - | 4 - 5 | 691 | 90 | ||
| BC003818.1 | - | 4 - 5 | 440 | 3489295 | 90 | |
| BC094053.1 | - | 4 - 5 | 681 | 3157927 | 90 | |
| EF460348.1 | - | 1 - 2 | 85 | 90 | ||
| M61909.1 | - | 4 - 5 | 396 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rela
Genomic Map