Assay Details
| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081079.1 | NP_001074548.1 | 3 - 4 | 527 | 101 | |
| GenBank mRNA | BC041673.1 | - | 2 - 3 | 1909 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogfrl1
Genomic Map