Assay Details
| Gene Symbol: Cnot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024327.1 | 2 - 3 | 204 | 116 | ||
| GenBank mRNA | AK011231.1 | - | 2 - 3 | 194 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
potassium large conductance calcium-activated channel, subfamily M, beta member 4, opposite strand 2 |
Kcnmb4os2 | 4933400F03Rik | -32.1kb | Anti-sense | Chr. 10 - 116473163 - 116474987 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 5330438D12 gene | 5330438D12Rik | 32.3kb | Anti-sense | Chr. 10 - 116581453 - 116583514 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011231.1 transcripts.
Set Membership: |
|

Cnot2
Genomic Map