Assay Details
| Gene Symbol: Fgf16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030614.2 | NP_085117.2 | 1 - 2 | 338 | 101 | |
| GenBank mRNA | AB049219.1 | - | 1 - 2 | 274 | 101 | |
| AF292104.1 | - | 1 - 2 | 274 | 101 | ||
| BC132641.1 | - | 1 - 2 | 338 | 40130918 | 101 | |
| BC137993.1 | - | 1 - 2 | 338 | 8861012 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgf16
Genomic Map