Assay Details
| Gene Symbol: Frs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138144.1 | - | 1 - 2 | 148 | 155 | |
| BC055334.1 | - | 1 - 2 | 124 | 6830555 | 155 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC055334.1 transcripts.
Set Membership: |
|

Frs2
Genomic Map