Assay Details
| Gene Symbol: Map2k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008928.4 | NP_032954.1 | 5 - 6 | 568 | 80 | |
| GenBank mRNA | AK008141.1 | - | 5 - 6 | 571 | 80 | |
| AK011002.1 | - | 5 - 6 | 569 | 80 | ||
| AK053378.1 | - | 5 - 6 | 569 | 80 | ||
| AK159379.1 | - | 5 - 6 | 567 | 80 | ||
| AK166255.1 | - | 5 - 6 | 570 | 80 | ||
| AK166507.1 | - | 5 - 6 | 569 | 80 | ||
| AK167209.1 | - | 5 - 6 | 569 | 80 | ||
| AK168710.1 | - | 5 - 6 | 551 | 80 | ||
| BC007467.1 | - | 5 - 6 | 559 | 3497714 | 80 | |
| D87115.1 | - | 5 - 6 | 581 | 80 | ||
| X93150.1 | - | 3 - 4 | 303 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k3
Genomic Map