Assay Details
| Gene Symbol: Map3k8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007746.2 | NP_031772.1 | 4 - 5 | 891 | 101 | |
| GenBank mRNA | AK140334.1 | - | 4 - 5 | 892 | 101 | |
| AK149624.1 | - | 4 - 5 | 892 | 101 | ||
| AK149846.1 | - | 4 - 5 | 892 | 101 | ||
| AK157123.1 | - | 4 - 5 | 858 | 101 | ||
| BC125286.1 | - | 3 - 4 | 792 | 40129801 | 101 | |
| BC137922.1 | - | 3 - 4 | 792 | 8860941 | 101 | |
| D13759.1 | - | 4 - 5 | 891 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k8
Genomic Map