Assay Details
| Gene Symbol: Cpsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013391.2 | NP_001013409.1 | 6 - 7 | 1275 | 105 | |
| GenBank mRNA | AK046856.1 | - | 6 - 7 | 1231 | 105 | |
| AK049749.1 | - | 6 - 7 | 1274 | 105 | ||
| AK077684.1 | - | 6 - 7 | 1277 | 105 | ||
| AK168764.1 | - | 6 - 7 | 1240 | 105 | ||
| BC068133.1 | - | 6 - 7 | 1251 | 6813785 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf6
Genomic Map