Assay Details
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133683.3 | NP_598444.1 | 4 - 5 | 616 | 77 | |
| GenBank mRNA | AK018383.1 | - | 4 - 5 | 564 | 77 | |
| AK034298.1 | - | 4 - 5 | 622 | 77 | ||
| AK036228.1 | - | 3 - 4 | 693 | 77 | ||
| AK042426.1 | - | 3 - 4 | 671 | 77 | ||
| AK079735.1 | - | 474 | 77 | |||
| AK136778.1 | - | 605 | 77 | |||
| AK138931.1 | - | 4 - 5 | 604 | 77 | ||
| AK155275.1 | - | 4 - 5 | 577 | 77 | ||
| AK170143.1 | - | 4 - 5 | 576 | 77 | ||
| AK171087.1 | - | 4 - 5 | 576 | 77 | ||
| AK193317.1 | - | 1 - 2 | 80 | 77 | ||
| BC016895.1 | - | 4 - 5 | 618 | 4239007 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem19
Genomic Map