Assay Details
| Gene Symbol: Slc22a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009203.3 | NP_033229.3 | 7 - 8 | 1600 | 60 | |
| GenBank mRNA | AB005451.1 | - | 7 - 8 | 1381 | 60 | |
| AK142580.1 | - | 6 - 7 | 749 | 60 | ||
| BC035927.1 | - | 1536 | 4242354 | 60 | ||
More Information
Set Membership: |
|

Slc22a12
Genomic Map