Assay Details
| Gene Symbol: Rhod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007485.4 | NP_031511.1 | 3 - 4 | 402 | 77 | |
| GenBank mRNA | AK008880.1 | - | 3 - 4 | 403 | 77 | |
| BC005652.1 | - | 3 - 4 | 399 | 77 | ||
| BC047989.1 | - | 3 - 4 | 411 | 6333308 | 77 | |
| D89821.1 | - | 413 | 77 | |||
| X84325.1 | - | 3 - 4 | 329 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rhod
Genomic Map