Assay Details
| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153502.3 | NP_705722.2 | 2 - 3 | 192 | 102 | |
| GenBank mRNA | AF492400.1 | - | 2 - 3 | 192 | 102 | |
| BC022973.1 | - | 351 | 102 | |||
| BC023221.1 | - | 2 - 3 | 169 | 102 | ||
| BC132370.1 | - | 2 - 3 | 202 | 40130647 | 102 | |
| BC138797.1 | - | 2 - 3 | 202 | 8861819 | 102 | |
| BC145421.1 | - | 2 - 3 | 202 | 9053979 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC022973.1 transcripts.
Set Membership: |
|

Ankrd23
Genomic Map