Assay Details
| Gene Symbol: Tram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028304.1 | - | 3 - 4 | 459 | 113 | |
| AK088814.1 | - | 3 - 4 | 450 | 113 | ||
| AK142658.1 | - | 3 - 4 | 450 | 113 | ||
| AK146207.1 | - | 3 - 4 | 449 | 113 | ||
| AK150551.1 | - | 3 - 4 | 456 | 113 | ||
| AK150777.1 | - | 3 - 4 | 414 | 113 | ||
| AK151240.1 | - | 3 - 4 | 413 | 113 | ||
| AK151409.1 | - | 3 - 4 | 456 | 113 | ||
| AK151664.1 | - | 3 - 4 | 456 | 113 | ||
| AK152338.1 | - | 3 - 4 | 456 | 113 | ||
| AK153379.1 | - | 3 - 4 | 456 | 113 | ||
| AK155905.1 | - | 3 - 4 | 450 | 113 | ||
| AK159203.1 | - | 3 - 4 | 450 | 113 | ||
| AK159350.1 | - | 3 - 4 | 450 | 113 | ||
| AK159878.1 | - | 3 - 4 | 459 | 113 | ||
| AK164347.1 | - | 3 - 4 | 362 | 113 | ||
| AK168708.1 | - | 3 - 4 | 392 | 113 | ||
| AY029764.1 | - | 3 - 4 | 342 | 113 | ||
| BC012401.1 | - | 3 - 4 | 439 | 3967323 | 113 | |
| CT010238.1 | - | 3 - 4 | 307 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tram1
Genomic Map