Assay Details
| Gene Symbol: E330032C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054493.1 | - | 1 - 2 | 163 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lysocardiolipin acyltransferase 1 | Lclat1 |
AI181996, Agpat8, Alcat1, Gm91, Lycat |
-9.2kb | Sense | Chr. 17 - 73107985 - 73243368 on NCBI Build GRCm38 |
| Downstream | calpain 13 | Capn13 |
CANP 13, Gm943 |
52.2kb | Anti-sense | Chr. 17 - 73306456 - 73399306 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

E330032C10Rik
Genomic Map