Assay Details
| Gene Symbol: Gm5089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033325.1 | 1 - 2 | 70 | 58 | ||
| GenBank mRNA | AK078338.1 | - | 1 - 2 | 72 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078338.1 transcripts.
Set Membership: |
|

Gm5089
Genomic Map