Assay Details
| Gene Symbol: Gm14858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040285.1 | 1 - 2 | 376 | 101 | ||
| GenBank mRNA | AK090266.1 | - | 1 - 2 | 377 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1 | Cited1 |
AI316840, AU019144, Msg1 |
-470.0 | Anti-sense | Chr. X - 102247378 - 102252181 on NCBI Build GRCm38 |
| Downstream | histone deacetylase 8 | Hdac8 | 2610007D20Rik | 31.0kb | Anti-sense | Chr. X - 102284640 - 102509686 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm14858
Genomic Map