Assay Details
| Gene Symbol: Gm34184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035016.1 | - | 1 - 2 | 169 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1500009L16 gene | 1500009L16Rik |
2700050C19Rik, OCC-1 |
-307.6kb | Anti-sense | Chr. 10 - 83722666 - 83762762 on NCBI Build GRCm38 |
| Downstream |
NUAK family, SNF1-like kinase, 1 |
Nuak1 |
AU014801, AW494241, B230104P22Rik, Omphk1 |
270.2kb | Sense | Chr. 10 - 84370900 - 84440471 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm34184
Genomic Map