Assay Details
| Gene Symbol: Mtpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026157.2 | NP_080433.1 | 1 - 2 | 190 | 81 | |
| GenBank mRNA | AK002680.1 | - | 1 - 2 | 180 | 81 | |
| AK011546.1 | - | 1 - 2 | 191 | 81 | ||
| AK036760.1 | - | 1 - 2 | 191 | 81 | ||
| AK045203.1 | - | 1 - 2 | 180 | 81 | ||
| AK076565.1 | - | 1 - 2 | 180 | 81 | ||
| AK135537.1 | - | 1 - 2 | 158 | 81 | ||
| AK154747.1 | - | 1 - 2 | 187 | 81 | ||
| AK166038.1 | - | 1 - 2 | 187 | 81 | ||
| BC017535.1 | - | 1 - 2 | 179 | 81 | ||
| BC057643.1 | - | 1 - 2 | 164 | 2616306 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtpap
Genomic Map