Assay Details
| Gene Symbol: Mtpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002680.1 | - | 2 - 3 | 347 | 105 | |
| AK166038.1 | - | 2 - 3 | 354 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtpap| Gene Symbol: Mtpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002680.1 | - | 2 - 3 | 347 | 105 | |
| AK166038.1 | - | 2 - 3 | 354 | 105 | ||
Set Membership: |
|