Assay Details
| Gene Symbol: Mtpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026157.2 | NP_080433.1 | 7 - 8 | 1344 | 141 | |
| GenBank mRNA | AK002680.1 | - | 7 - 8 | 1315 | 141 | |
| AK011546.1 | - | 7 - 8 | 1345 | 141 | ||
| AK036760.1 | - | 7 - 8 | 1349 | 141 | ||
| AK076565.1 | - | 7 - 8 | 1334 | 141 | ||
| AK135537.1 | - | 7 - 8 | 1312 | 141 | ||
| AK154747.1 | - | 7 - 8 | 1345 | 141 | ||
| AK166038.1 | - | 7 - 8 | 1322 | 141 | ||
| BC017535.1 | - | 7 - 8 | 1337 | 141 | ||
| BC057643.1 | - | 7 - 8 | 1318 | 2616306 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtpap
Genomic Map