Assay Details
| Gene Symbol: Gm5089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047660.1 | - | 2 - 3 | 245 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | citrate lyase beta like | Clybl |
0610033J05Rik, 2310014M14Rik, AI256068, Clb |
Anti-sense | Chr. 14 - 122171283 - 122402234 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein of the cerebellum 5 | Zic5 |
1700049L20Rik, Opr |
62.5kb | Sense | Chr. 14 - 122459160 - 122465716 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm5089
Genomic Map