Assay Details
| Gene Symbol: Gm5089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033326.1 | 3 - 4 | 221 | 74 | ||
| GenBank mRNA | AK047660.1 | - | 4 - 5 | 468 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | citrate lyase beta like | Clybl |
0610033J05Rik, 2310014M14Rik, AI256068, Clb |
Anti-sense | Chr. 14 - 122171283 - 122402234 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein of the cerebellum 5 | Zic5 |
1700049L20Rik, Opr |
90.8kb | Sense | Chr. 14 - 122459160 - 122465716 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm5089
Genomic Map