Assay Details
| Gene Symbol: Dzip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028258.3 | NP_082534.2 | 11 - 12 | 1573 | 75 | |
| GenBank mRNA | AK028787.1 | - | 12 - 13 | 1728 | 75 | |
| AK034503.1 | - | 11 - 12 | 1619 | 75 | ||
| AK166083.1 | - | 12 - 13 | 1732 | 75 | ||
| BC099950.1 | - | 11 - 12 | 1573 | 30094307 | 75 | |
| BC138218.1 | - | 10 - 11 | 1502 | 8861237 | 75 | |
| BC145204.1 | - | 10 - 11 | 1502 | 9053755 | 75 | |
More Information
Set Membership: |
|

Dzip1l
Genomic Map