Assay Details
| Gene Symbol: Dzip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028258.3 | NP_082534.2 | 6 - 7 | 1145 | 63 | |
| GenBank mRNA | AK028787.1 | - | 7 - 8 | 1300 | 63 | |
| AK034503.1 | - | 6 - 7 | 1191 | 63 | ||
| AK166083.1 | - | 7 - 8 | 1304 | 63 | ||
| AK215157.1 | - | 1 - 2 | 69 | 63 | ||
| BC099950.1 | - | 6 - 7 | 1145 | 30094307 | 63 | |
| BC138218.1 | - | 5 - 6 | 1074 | 8861237 | 63 | |
| BC145204.1 | - | 5 - 6 | 1074 | 9053755 | 63 | |
More Information
Set Membership: |
|

Dzip1l
Genomic Map