Assay Details
| Gene Symbol: Dzip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028258.3 | NP_082534.2 | 8 - 9 | 1347 | 78 | |
| GenBank mRNA | AK028787.1 | - | 9 - 10 | 1502 | 78 | |
| AK034503.1 | - | 8 - 9 | 1393 | 78 | ||
| AK166083.1 | - | 9 - 10 | 1506 | 78 | ||
| AK215157.1 | - | 3 - 4 | 271 | 78 | ||
| BC099950.1 | - | 8 - 9 | 1347 | 30094307 | 78 | |
| BC138218.1 | - | 7 - 8 | 1276 | 8861237 | 78 | |
| BC145204.1 | - | 7 - 8 | 9053755 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dzip1l
Genomic Map