Assay Details
| Gene Symbol: Gpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175500.4 | NP_780709.1 | 3 - 4 | 1433 | 64 | |
| GenBank mRNA | AF001463.1 | - | 1 - 2 | 83 | 64 | |
| AK038547.1 | - | 3 - 4 | 1398 | 64 | ||
| AK084223.1 | - | 3 - 4 | 1377 | 64 | ||
| BC115712.1 | - | 3 - 4 | 1225 | 40059645 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpc5
Genomic Map