Assay Details
| Gene Symbol: Gpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175500.4 | NP_780709.1 | 5 - 6 | 1690 | 74 | |
| GenBank mRNA | AK038547.1 | - | 5 - 6 | 1655 | 74 | |
| BC115712.1 | - | 5 - 6 | 1482 | 40059645 | 74 | |
More Information
Set Membership: |
|

Gpc5
Genomic Map