Assay Details
| Gene Symbol: Gm29506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046640.1 | - | 67 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4933415F23 gene | 4933415F23Rik | -112.4kb | Sense | Chr. 1 - 23100473 - 23102253 on NCBI Build GRCm38 | |
| Downstream | microRNA 30a | Mir30a |
Mirn30a, mir-30a, mmu-mir-30a |
36.6kb | Anti-sense | Chr. 1 - 23272269 - 23272339 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm29506
Genomic Map