Assay Details
| Gene Symbol: Foxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180974.4 | NP_851305.2 | 1 - 2 | 171 | 88 | |
| GenBank mRNA | AK030735.1 | - | 1 - 2 | 126 | 88 | |
| AK031639.1 | - | 1 - 2 | 109 | 88 | ||
| AK154401.1 | - | 1 - 2 | 163 | 88 | ||
| BC138824.1 | - | 1 - 2 | 58 | 88 | ||
| BC138825.1 | - | 1 - 2 | 58 | 88 | ||
| CN698353.1 | - | 1 - 2 | 102 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030735.1,BC138824.1,BC138825.1,NM_180974.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Foxn2
Genomic Map