Assay Details
| Gene Symbol: Ppp1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028658.4 | NP_082934.3 | 15 - 16 | 1733 | 82 | |
| GenBank mRNA | AK155094.1 | - | 15 - 16 | 1565 | 82 | |
| AK170254.1 | - | 15 - 16 | 1621 | 82 | ||
| BC137673.1 | - | 15 - 16 | 1673 | 8860692 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r21
Genomic Map