Assay Details
| Gene Symbol: 4930538E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019722.1 | - | 2 - 3 | 310 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | poly(A) polymerase gamma | Papolg | 9630006B20Rik | -271.6kb | Sense | Chr. 11 - 23860145 - 23895296 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4933430M04 gene | 4933430M04Rik | 313.1kb | Anti-sense | Chr. 11 - 24481909 - 24569951 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930538E20Rik
Genomic Map